Learn More
OriGene Flp Rabbit Polyclonal Antibody, 100 uL
SDP
Supplier:  OriGene TA160030
Encompass_Preferred
Flp recombinase originates from Saccharomyces cerevisiae and recognizes FRT sites in the genome. It catalyzes recombination between two FRT sites, thereby excising the intervening DNA sequence. Flp recombinase is used as a tool for the generation of transgenic animals.
Catalog No. NC1221827