Learn More
Bellco Glass, Inc. Anti-Foam Assembly , 15L
SDP
Catalog No. 501946493
Encompass_Preferred
Adjustable anti-foam probe works by conductance. Rising level in bioreactor contacts the probe to trigger the controller for addition of anti-foam or addition / withdraw of media to maintain proper liquid level. Fits into 18 mm port on 3L metric head plate. Supplied with nut, washers and O-Rings. MADE IN THE USA
Catalog No. 50-194-6493 Supplier Bellco Glass, Inc. Supplier No. 790915111
Adjustable anti-foam probe works by conductance. Rising level in bioreactor contacts the probe to trigger the controller for addition of anti-foam or addition / withdraw of media to maintain proper liquid level. Fits into 18 mm port on 3L metric head plate. Supplied with nut, washers and O-Rings. MADE IN THE USA