Learn More
Bellco Glass, Inc. Anti-Foam Assembly , 3L For 18mm Port
SDP
Catalog No. 501946460
Encompass_Preferred
Anti Foam Level Probe Assembly Adjustable anti-foam probe works by conductance. Rising level in bioreactor contacts the probe to trigger the controller for addition of anit-foam or addition / withdraw of media to maintain proper liquid level. Fits into 18 mm port on 3L metric head plate. Supplied with nut, washers and O-Rings.. MADE IN THE USA
Catalog No. 50-194-6460 Supplier Bellco Glass, Inc. Supplier No. 790900086
Anti Foam Level Probe Assembly Adjustable anti-foam probe works by conductance. Rising level in bioreactor contacts the probe to trigger the controller for addition of anit-foam or addition / withdraw of media to maintain proper liquid level. Fits into 18 mm port on 3L metric head plate. Supplied with nut, washers and O-Rings.. MADE IN THE USA