Learn More
Uline Anti-Fog Cleaning Spray, 8 oz, ****Minimum order quantity of 2 each****
Catalog No. NC3311922
Encompass_Preferred
Clean and protect your face shields, goggles and glasses.Spray on and wipe off.Keeps lenses fog-free for up to 8 hours.****Minimum order quantity of 2 each****
Catalog No. NC3311922 Supplier Uline Supplier No. S25645EA
Clean and protect your face shields, goggles and glasses.Spray on and wipe off.Keeps lenses fog-free for up to 8 hours.****Minimum order quantity of 2 each****