Learn More
Grainger Approved Vendor Fuse Series Safety Glasses, Clear Lens, Frameless, 1 ea
Catalog No. NC1158090
Encompass_Preferred
These Fuse series safety glasses offer reliable eye protection for a wide range of applications. Designed with a lightweight, frameless construction and a wraparound style, they provide excellent peripheral vision and a comfortable fit.
  • Clear polycarbonate lens construction
  • Clear frame and arm design
  • Wraparound frame style for increased coverage
  • Universal eyewear size suitable for most users
  • Straight arm tip style
  • Meets ANSI Z87.1-2015 standards
This item is not returnable. View return policy
Catalog No. NC1158090 Supplier Grainger Supplier No. 34CT37
These Fuse series safety glasses offer reliable eye protection for a wide range of applications. Designed with a lightweight, frameless construction and a wraparound style, they provide excellent peripheral vision and a comfortable fit.
  • Clear polycarbonate lens construction
  • Clear frame and arm design
  • Wraparound frame style for increased coverage
  • Universal eyewear size suitable for most users
  • Straight arm tip style
  • Meets ANSI Z87.1-2015 standards