Learn More
Poltex Anti-Fog Surgical Mask Holder-2 Visual Management
SDP
Catalog No. 22000470
Change view
Click to view available options
Mounting:
Magnetic Mount
Wall Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-000-470 Wall Mount
22-000-469 Magnetic Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Consumables shown for demonstration purposes only and are not included with the product.
Catalog No. 22-000-470 Supplier Poltex Supplier No. ANTIFOG2VMW
  • $135.00
    $126.65 / Each of 1
    Save $8.35
    6% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Consumables shown for demonstration purposes only and are not included with the product.
Visually manage two boxes of anti-fog surgical masks.
  • Constructed from super-durable PETG
  • When lower box is empty, front face lifts up and new box falls into place
  • Mounts with powerful magnets or keyhole slots for wall-mount
  • Consumables not included; for demonstration purposes only
  • See image for dimensions
  • Hardware not included for wall mount unit (-W). Customer must provide their own screw/anchor for their wall type.
Specifications
Product Type Face Mask Dispenser
Material PETG
For Use With (Application) Healthcare
For Use With (Equipment) Surgical masks
Mounting Wall Mount