Learn More
Immunology Consultants anti-Foot-And-Mouth Disease Antibody (Mouse - Clone #100) - Monoclonal
SDP
Catalog No. 5000435290
Encompass_Preferred
This monoclonal antibody provides high‑specificity detection of Foot‑and‑Mouth Disease Virus (FMDV) VP2 antigen, supporting ELISA, Western blotting, and antigen‑capture assays. Clone 100 delivers strong affinity and low background, enabling sensitive detection in clinical samples, culture supernatants, and purified antigen preparations. MFMV‑55A‑100 is ideal for virology research, vaccine evaluation, and surveillance of FMDV infection in livestock. Its consistent performance ensures reliable antigen detection across repeated assays.
Catalog No. 50-004-35290 Supplier Immunology Consultants Supplier No. MFMV55A100
This monoclonal antibody provides high‑specificity detection of Foot‑and‑Mouth Disease Virus (FMDV) VP2 antigen, supporting ELISA, Western blotting, and antigen‑capture assays. Clone 100 delivers strong affinity and low background, enabling sensitive detection in clinical samples, culture supernatants, and purified antigen preparations. MFMV‑55A‑100 is ideal for virology research, vaccine evaluation, and surveillance of FMDV infection in livestock. Its consistent performance ensures reliable antigen detection across repeated assays.