Learn More
Sino Biological Anti-FXN/Frataxin Antibody, Rabbit Polyclonal 100
Catalog No. 5000750134
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human FXN, and purified by antigen affinity chromatography.
Catalog No. 50-007-50134 Supplier Sino Biological Supplier No. 102315T40100
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human FXN, and purified by antigen affinity chromatography.