Learn More
ECM Bioscience FXYD5/Dysadherin (Extracellular region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience FM0291
Encompass_Preferred
Clone (M029) was generated from a proprietary antigen related to the native human FXYD5 protein expressed in A431 epidermoid carcinoma cell line. Accession #Q96DB9. Clone M029 mouse monoclonal antibody detects a 35-55 kDa* band on SDS-PAGE immunoblots of human A431, A549, MeWo, MDA-MB-231 cell, as well as human breast, lung, and skin tissues. The antibody works for western blot, immunoprecipitation, ELISA, and immunocytochemistry, as well as detects the FXYD5 protein on live cells. Applications: WB, E, IP, ICC; Species Reactivity: Hu; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein G chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = FXYD domain-containing ion transport regulator 5, Dysadherin, FXYD5, DYSAD, IWU1, HSPC113, UNQ2561/PRO6241
Catalog No. 50-206-8715