Learn More
Cell Signaling Technology G4S Linker Rabbit mAb, Pacific Blue Conjugate, Flow Cytometric Analysis, 15.84
SDP
Supplier:  Cell Signaling Technology 44962S
Encompass
The G4S Linker (E7O2V) Rabbit Monoclonal Antibody (Pacific Blue™ Conjugate) enables precise detection of CARs with a G4S linker, enhancing flow cytometric analysis of live cells. This recombinant antibody offers high specificity and robustness, critical for stability and performance in experiments involving engineered receptors, setting it apart in antibody-based applications.
Catalog No. NC2883617