Learn More
Sino Biological Anti-GAD65/GAD2 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000785320
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse GAD65/GAD2 (rM GAD65/GAD2; Catalog#50653-MNCB; NP_032104.2; Met1-Leu585). GAD65/GAD2 specific IgG was purified by Mouse GAD65/GAD2 affinity chromatography.
Catalog No. 50-007-85320 Supplier Sino Biological Supplier No. 50653T60100
Produced in rabbits immunized with purified, recombinant Mouse GAD65/GAD2 (rM GAD65/GAD2; Catalog#50653-MNCB; NP_032104.2; Met1-Leu585). GAD65/GAD2 specific IgG was purified by Mouse GAD65/GAD2 affinity chromatography.