Learn More
Ubpbio ANTI-GAPDH, MOUSE 100UG
Supplier:  Ubpbio Y1041
Encompass_Preferred
Antibodies – Anti-GAPDH (UBPBio catalog# Y1041) – Mouse monoclonal antibody (clone GA1R, IgG1). Used for immunoblotting at 1:1,000-10,000 dilution for detection of GAPDH in 20 ug whole cell lysates. Recognizing GAPDH in many species including E. coli, yeast, insects, human, mouse, rat, chicken, rabbit and hamster.
Catalog No. 50-112-6919