Learn More
Amsbio LLC Anti-Gb3 Monoclonal Antibody
Supplier:  Amsbio LLC AMS.A2506
Encompass
Clone Name: BGR23; Specificity: Glycolipid Gb3; Immunogen: Purified glycolipid Gb3 adsorbed to Salmonella minnesota; Applications: TLC-immunostainig, ELISA, Immunohistochemistry
Catalog No. 50-204-1196