Learn More
Biolegend Anti-GFAP
Supplier:  Biolegend 837401
Encompass_Preferred
Anti-GFAP [SMI 24]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P, WB; Size: 100 uL
Catalog No. 50-207-4398