Learn More
Biolegend Anti-GFAP
Supplier:  Biolegend 837202
Encompass_Preferred
Anti-GFAP [SMI 21]; Isotype: Mouse IgG1, κ; Reactivity: Human, Monkey, Canine; Apps: IHC-P, WB, ICC; Size: 500 uL
Catalog No. 50-103-0009