Learn More
STEMCELL Technologies Anti-GFAP Antibody, Clone 2E1.E9, Size: 25 µg
SDP
Supplier:  STEMCELL Technologies 60048.1
Encompass
Mouse monoclonal IgG2b antibody against human, mouse, rat GFAP (glial fibriallary acidic protein), unconjugated
Catalog No. 50-197-5311
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov