Learn More
Immunology Consultants anti-GFAP Antibody - Monoclonal (Clone #100)
SDP
Supplier:  Immunology Consultants MGFAP80A100
Encompass_Preferred
This monoclonal antibody provides highly specific detection of Glial Fibrillary Acidic Protein (GFAP), a key intermediate filament protein expressed in astrocytes and widely used as a marker of neural injury, gliosis, and neurodegenerative processes. Clone 100 delivers strong affinity and low background in ELISA, Western blotting, immunocytochemistry, and tissue‑based assays. Its clean, reproducible signal supports research on CNS pathology, astrocyte activation, and biomarker development. MGFAP‑80A‑100 performs reliably across diverse sample types, enabling precise GFAP quantification in both basic and translational neuroscience workflows
Catalog No. 50-004-35476