Learn More
STEMCELL Technologies Anti-GFAP Antibody, Polyclonal, Size: 200 µL
SDP
Supplier:  STEMCELL Technologies 60128
Encompass
This rabbit polyclonal antibody reacts with glial fibrillary acidic protein (GFAP), an ~55 kDa type III intermediate filament protein. Verified for use with immunohistochemistry and western blotting assays, use this antibody in applications requiring the detection of GFAP. GFAP is a differentiation marker for astrocytes, and it is localized in normal central nervous system tissue, and certain tumor and metastases of the glial antigen. This antibody is suitable for immunofluorescent labeling of cultured mammalian cells. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5703