Learn More
Biolegend Anti-GFP Nanobody Affinity Gel
Supplier:  Biolegend 689302
Encompass_Preferred
Anti-GFP Nanobody Affinity Gel [LaG-19]; Isotype: Llama VH Ig; Reactivity: Aequorea victoria, Epitope tag; Apps: IP; Size: 1 mL
Catalog No. 50-272-1767