Learn More
Immunology Consultants anti-Glu-Glu Antibody (Rabbit) - Affinity Purified
SDP
Catalog No. 5000435351
Encompass_Preferred
RGLU‑45A is an affinity‑purified rabbit polyclonal antibody specific for the Glu‑Glu tag, a short epitope used in recombinant protein expression and detection. The purification process enriches high‑affinity antibodies, ensuring strong specificity and reduced background in Western blotting, ELISA, immunoprecipitation, and immunofluorescence applications. Produced by Immunology Consultants and supplied in a 1 mg format, RGLU‑45A provides a high‑quality reagent for researchers working with Glu‑Glu–tagged proteins.
Catalog No. 50-004-35351 Supplier Immunology Consultants Supplier No. RGLU45A
RGLU‑45A is an affinity‑purified rabbit polyclonal antibody specific for the Glu‑Glu tag, a short epitope used in recombinant protein expression and detection. The purification process enriches high‑affinity antibodies, ensuring strong specificity and reduced background in Western blotting, ELISA, immunoprecipitation, and immunofluorescence applications. Produced by Immunology Consultants and supplied in a 1 mg format, RGLU‑45A provides a high‑quality reagent for researchers working with Glu‑Glu–tagged proteins.