Learn More
Amsbio LLC Anti-GM3 Monoclonal Antibody
Catalog No. 502041202
Encompass
Clone Name: GMR6; Specificity: Ganglioside GM3; Immunogen: Purified ganglioside GM3 adsorbed to Salmonella minnesota; Applications: TLC-immunostaining, Flow cytometric analysis, ELISA, Immunohistochemistry
Catalog No. 50-204-1202 Supplier Amsbio LLC Supplier No. AMS.A2582
Add to Cart
Add to Cart
Clone Name: GMR6; Specificity: Ganglioside GM3; Immunogen: Purified ganglioside GM3 adsorbed to Salmonella minnesota; Applications: TLC-immunostaining, Flow cytometric analysis, ELISA, Immunohistochemistry