Learn More
ABclonal Technology GNMT Polyclonal Antibody, 100UL, Validated in:WB
SDP
Supplier:  ABclonal Technology A6608
Encompass_Preferred
Immunogen: Recombinant fusion protein containing a sequence corresponding to amino acids 1-295 of human GNMT (NP_061833.1).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Mouse,Rat•Validated Applications:WB•Swiss Prot: Q14749•Gene ID:27232•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-130-8538