Learn More
Seracare Life Sciences Inc ANTI-GOAT IGG (H+L) AB BIOTIN
Supplier: Seracare Life Sciences Inc 52600035
Affinity purified polyclonal antibody to Goat IgG, heavy and light chains (whole IgG) made in Rabbit and biotinylated. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.