Learn More
Seracare Life Sciences Inc ANTI-GOAT IGG (H+L) AB BIOTIN
Catalog No. 5067574
Encompass_Preferred
Affinity purified polyclonal antibody to Goat IgG heavy and light chains (whole IgG) made in Rabbit and biotinylated. This product is specifically for use in immunohistochemistry assays and is provided as a liquid at use dilution. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable. View return policy
Catalog No. 50-675-74 Supplier Seracare Life Sciences Inc Supplier No. 55700009
Affinity purified polyclonal antibody to Goat IgG heavy and light chains (whole IgG) made in Rabbit and biotinylated. This product is specifically for use in immunohistochemistry assays and is provided as a liquid at use dilution. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.