Learn More
Seracare Life Sciences Inc ANTI-GOAT IGG (H+L) AB CY5
Catalog No. 5067058
Encompass_Preferred
Affinity purified polyclonal antibody to Goat IgG, both heavy and light chains (whole IgG) made in Rabbit and labeled with Cy5. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
This item is not returnable. View return policy
Catalog No. 50-670-58 Supplier Seracare Life Sciences Inc Supplier No. 52300371
Affinity purified polyclonal antibody to Goat IgG, both heavy and light chains (whole IgG) made in Rabbit and labeled with Cy5. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).