Learn More
Seracare Life Sciences Inc ANTI-GOAT IGG (H+L) AB DL488
Catalog No. 5067077
Encompass_Preferred
Affinity purified polyclonal antibody to Goat IgG, both heavy and light chains (whole IgG) made in Rabbit and labeled with DyLight 488. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
This item is not returnable. View return policy
Catalog No. 50-670-77 Supplier Seracare Life Sciences Inc Supplier No. 52300383
Affinity purified polyclonal antibody to Goat IgG, both heavy and light chains (whole IgG) made in Rabbit and labeled with DyLight 488. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).