Learn More
Seracare Life Sciences Inc ANTI-GOAT IGG (H+L) AB FITC
Catalog No. 5066880
Encompass_Preferred
Affinity purified polyclonal antibody to Goat IgG, both heavy and light chains (whole IgG) made in Rabbit and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
This item is not returnable. View return policy
Catalog No. 50-668-80 Supplier Seracare Life Sciences Inc Supplier No. 52300294
Affinity purified polyclonal antibody to Goat IgG, both heavy and light chains (whole IgG) made in Rabbit and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.