Learn More
Rockland Immunochemicals Anti-Goat IgG [H&L] HRP Conjugated Pre-Adsorbed 1 mg
SDP
Catalog No. 501943872
Encompass_Preferred
Anti-Goat IgG generated in rabbit detects goat Immunoglobulin G. It is a protein complex composed of four peptide chains — two identical heavy chains and two identical light chains arranged in a Y-shape typical of antibody monomers. Each IgG has two antigen binding sites. Representing approximately 75% of serum immunoglobulins IgG is the most abundant antibody isotype found in the circulation. IgG molecules are synthesized and secreted by plasma B cells. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition. Anti-Goat IgG Antibody Pre-Adsorbed is conjugated to horseradish peroxidase.
Catalog No. 50-194-3872 Supplier Rockland Immunochemicals Supplier No. 605403B69
Anti-Goat IgG generated in rabbit detects goat Immunoglobulin G. It is a protein complex composed of four peptide chains — two identical heavy chains and two identical light chains arranged in a Y-shape typical of antibody monomers. Each IgG has two antigen binding sites. Representing approximately 75% of serum immunoglobulins IgG is the most abundant antibody isotype found in the circulation. IgG molecules are synthesized and secreted by plasma B cells. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition. Anti-Goat IgG Antibody Pre-Adsorbed is conjugated to horseradish peroxidase.