Learn More
Rockland Immunochemicals TrueBlot Anti-Goat Ig IP Agarose Beads - 00-8844-25
SDP
Catalog No. NC9277501
Encompass_Preferred
TrueBlot Anti-Goat Ig IP Agarose Beads are a suspension of activated agarose beads coupled with rabbit Anti-goat IgG. It is suitable for precipitation of goat IgGs used as the primary antibodies in immunoprecipitation assays. The beads are in suspension and will settle upon storage. Prior to use, mix the vial gently (do not vortex) to ensure delivery of proper bead volume.
Catalog No. NC9277501 Supplier Rockland Immunochemicals Supplier No. 00884425
TrueBlot Anti-Goat Ig IP Agarose Beads are a suspension of activated agarose beads coupled with rabbit Anti-goat IgG. It is suitable for precipitation of goat IgGs used as the primary antibodies in immunoprecipitation assays. The beads are in suspension and will settle upon storage. Prior to use, mix the vial gently (do not vortex) to ensure delivery of proper bead volume.