Learn More
ECM Bioscience GSK-3β (N-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience GM1311
Encompass_Preferred
Clone M131 was generated from a recombinant protein containing amino acid residues in the N-terminal region of human GSK-3β. This sequence is highly conserved in rat and mouse GSK-3β, and has 65% homology to similar regions in GSK-3α. Accession #P49841. This mouse monoclonal antibody was purified with protein A chromatography. The antibody detects a 46 kDa* protein corresponding to the apparent molecular mass of GSK-3β on SDS-PAGE immunoblots of rabbit fibroblasts, mouse brain, as well as human SKN-SH and A431 cells. Applications: WB, E, IHC, ICC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Glycogen synthase kinasebeta3
Catalog No. 50-206-8720