Learn More
BIOHIPPO Anti-GST 26 Nanobody (ABS0324), 100 µg
SDP
Catalog No. 5000860121
Encompass_Preferred
GST 26 Nanobody Recombinant Antibody (clone ABS0324) is a recombinant Mouse VHH, mFc antibody specific for GST 26 Nanobody. Suitable for ELISA, IF, SPR, WB. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-60121 Supplier BIOHIPPO Supplier No. BHA21405168
GST 26 Nanobody Recombinant Antibody (clone ABS0324) is a recombinant Mouse VHH, mFc antibody specific for GST 26 Nanobody. Suitable for ELISA, IF, SPR, WB. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.