Learn More
Seracare Life Sciences Inc ANTI-GUINEAPIGIGG(H+L)AB DL488
Supplier:  Seracare Life Sciences Inc 52300388
Encompass_Preferred
Affinity purified polyclonal antibody to Guinea Pig IgG, both heavy and light chains (whole IgG) made in Goat and labeled with DyLight 488. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-670-82