Learn More
Seracare Life Sciences Inc ANTI-GUINPIG IGG(H+L) AB DL549
Catalog No. 5067107
Encompass_Preferred
Affinity purified polyclonal antibody to Guinea Pig IgG, both heavy and light chains (whole IgG) made in Goat and labeled with DyLight 549. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
This item is not returnable. View return policy
Catalog No. 50-671-07 Supplier Seracare Life Sciences Inc Supplier No. 52300397
Affinity purified polyclonal antibody to Guinea Pig IgG, both heavy and light chains (whole IgG) made in Goat and labeled with DyLight 549. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).