Learn More
Bio X Cell anti-human CD1a, Clone OKT-6, IgG1, 5 MG
Supplier: Bio X Cell BE0211R005MG
The OKT-6 monoclonal antibody reacts with human CD1a, a member of the CD1 family of transmembrane glycoproteins. The CD1 proteins are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with β2-microglobulin. These proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells.
- Clone: OKT-6
- Target: Human CD1a
- Host: Mouse
- Isotype: Mouse IgG1
- Applications: In vitro CD1a blockade, flow cytometry
- Purification method: Protein G
- Formulation: PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin level: ≤1EU/mg (≤0.001EU/μg) by LAL assay
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.