Learn More
Bio X Cell Anti-Human CD3 UCHT1, Mouse IgG1, 5 MG
Supplier: Bio X Cell BE0231R005MG
The UCHT1 (Leu-4)(T3) monoclonal antibody reacts with human CD3ε, a 20 kDa transmembrane cell-surface protein of the immunoglobulin superfamily. CD3ε forms part of the TCR complex and is expressed on T lymphocytes, NK-T cells, and developing thymocytes. CD3 plays roles in TCR signaling, T lymphocyte activation, and antigen recognition.
- Clone: UCHT1 (Leu-4) (T3)
- Isotype: Mouse IgG1, κ
- Reactivity: Human
- Immunogen: Human CD3ε
- Purity: ≥95% by SDS-PAGE
- Endotoxin: ≤1EU/mg
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Sterility: 0.2 μm filtered
- Molecular Weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.