Learn More
Bio X Cell Antibody, CD3, OKT-3, IgG2a Unconjugated, 5 MG
Supplier: Bio X Cell BE00012R005MG
The OKT-3 monoclonal antibody reacts with human CD3ε, a 20 kDa transmembrane cell-surface protein belonging to the immunoglobulin superfamily. It plays roles in TCR signaling, T lymphocyte activation, and antigen recognition. The OKT-3 antibody has immunosuppressive properties in vivo and has been shown to effectively treat renal, heart, and liver allograft rejection.
- Isotype: Mouse IgG2a, κ
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Purity: ≥95% by SDS-PAGE
- Endotoxin: ≤1 EU/mg
- Sterility: 0.2 μm filtered
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Applications: In vitro T cell stimulation/activation, in vivo T cell depletion in humanized mice, ex vivo T cell inhibition for xenografts, Flow cytometry, in vitro Organoids/Organ-on-Chip
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.