Learn More
Bio X Cell Anti-Human CD47, B6.H12, Mouse IgG1, 25 MG
Supplier: Bio X Cell BE00191R025MG
The B6H12 monoclonal antibody reacts with human CD47, a ≈50 kDa glycosylated transmembrane protein ubiquitously expressed by hematopoietic and non-hematopoietic cells. Involved in apoptosis, proliferation, adhesion, and migration, it plays a role in immune and angiogenic responses. B6H12 neutralizes CD47 and inhibits growth of hepatocellular carcinoma cells in vitro.
- Applications: in vitro and in vivo CD47 neutralization, flow cytometry
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%, determined by SDS-PAGE
- Sterility: 0.2 μm filtration
- Production: purified from cell culture supernatant in animal-free facility
- Purification method: Protein G
- Storage: store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.