Learn More
Seracare Life Sciences Inc ANTI-H PYLORI ANTIBODY
Supplier:  Seracare Life Sciences Inc 53100325
Encompass_Preferred
Affinity purified polyclonal antibody to Helicobacter pylori made in Goat. Isolated from a serum pool of goats immunized with heat-killed whole cells of Helicobacter pylori. This antibody reacts with H. pylori 43504, 43526, and 43579 from ATCC.
Catalog No. 50-668-62