Learn More
REVMAB BIOSCIENCES USA INC Histone H3.3G34R [RM240], Rabbit Monoclonal Antibody
Supplier:  REVMAB BIOSCIENCES USA INC 31112000S
Encompass_Preferred
Anti-H3.3G34R Mutant rabbit monoclonal antibody, CloneRM240 (100uL); WB, IHC validated; Product info: www.revmab.com
Catalog No. NC1728981