Learn More
REVMAB BIOSCIENCES USA INC Histone H3.3 G34V [RM307], Rabbit Monoclonal Antibody
Supplier:  REVMAB BIOSCIENCES USA INC 31119300S
Encompass_Preferred
Anti-Histone H3.3 G34V Mutant Rabbit Monoclonal Antibody, CloneRM307 (100uL); WB, ELISA, IHC, ICC validated; Product info: www.revmab.com
Catalog No. NC1728984