Learn More
Biolegend Anti-HA.11 Epitope Tag
Supplier:  Biolegend 901513
Encompass_Preferred
Anti-HA.11 Epitope Tag [16B12]; Isotype: Mouse IgG1, κ; Reactivity: YPYDVPDYA Tag; Apps: WB, ICC, IP, FC, Purification; Size: 200 uL
Catalog No. 50-103-0107