Learn More
Bio X Cell Monoclonal Antibody, anti-HA, Mouse IgG2b, 1 MG
Supplier: Bio X Cell RT0268R001MG
The 12CA5 monoclonal antibody recognizes the 9-amino acid sequence YPYDVPDYA, which is derived from influenza virus hemagglutinin (HA). The HA tag is commonly added to proteins of interest using recombinant DNA technology and can then be used for detection or purification of the tagged protein.
- Isotype: Mouse IgG2b
- Conjugation: Unconjugated
- Immunogen: Influenza virus hemagglutinin protein (AA 98-106)
- Reported applications: ELISA, Western blot, Immunoprecipitation, Immunofluorescence
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Purity: ≥95%, determined by SDS-PAGE
- Endotoxin: ≤1 EU/mg
- Molecular weight: 150 kDa
- Storage: Store at 4°C; do not freeze
- Purification method: Protein G
- Production: Purified from cell culture supernatant in an animal-free facility
- Sterility: 0.2 μm filtration
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.