Learn More
Aves Labs, Inc Anti-HA Epitope Tag Antibody
Supplier:  Aves Labs, Inc ETHA100
Encompass
Clonality: Polyclonal | Form: Affinity Purified | Host Species: Chicken | Applications: ELISA, ICC, IHC, IP, WB
Catalog No. NC1566846