Learn More
Sigma Aldrich Fine Chemicals Biosciences ANTI-HA HIGH AFFINITY,RAT,50UG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11867423001
Encompass
Anti-HA High Affinity 3F10 Rat AntibodyAnti-HA High Affinity is a monoclonal antibody to the HA-peptide (clone 3F10). Anti-HA High Affinity recognizes the HA peptide sequence (YPYDVPDYA), derived from the influenza hemagglutinin protein. The antibody recognizes its antigenic determinant even when the HA peptide epitope is introduced into unrelated recombinant proteins by a technique known as "epitope tagging".
Catalog No. NC1821908