Learn More
Immunology Consultants anti-Hamster (CHO) Beta 2-Microglobulin Antibody - Affinity Purified
SDP
Catalog No. 5000435372
Encompass_Preferred
This affinity‑purified anti‑Hamster (CHO) Beta‑2‑Microglobulin (β2M) antibody provides a high‑specificity reagent for detecting this key component of MHC class I molecules in CHO‑derived systems. β2M is a widely monitored host cell protein due to its role in antigen presentation and its potential presence as a process‑related impurity in CHO‑based biologics manufacturing. Supplied at 0.1 mg, this antibody delivers consistent performance in ELISA, Western blotting, and other immunoassay formats. Its high affinity and CHO‑targeted specificity make it an excellent tool for HCP profiling, purification validation, and analytical method development where sensitive detection of β2M is required.
Catalog No. 50-004-35372 Supplier Immunology Consultants Supplier No. CB265ALYZ
This affinity‑purified anti‑Hamster (CHO) Beta‑2‑Microglobulin (β2M) antibody provides a high‑specificity reagent for detecting this key component of MHC class I molecules in CHO‑derived systems. β2M is a widely monitored host cell protein due to its role in antigen presentation and its potential presence as a process‑related impurity in CHO‑based biologics manufacturing. Supplied at 0.1 mg, this antibody delivers consistent performance in ELISA, Western blotting, and other immunoassay formats. Its high affinity and CHO‑targeted specificity make it an excellent tool for HCP profiling, purification validation, and analytical method development where sensitive detection of β2M is required.