Learn More
Immunology Consultants anti-Hamster (CHO) C-C Motif Chemokine Antibody - Affinity Purified
SDP
Catalog No. 5000435325
Encompass_Preferred
This affinity‑purified goat antibody provides highly specific detection of CHO‑derived C‑C Motif Chemokine, an important host cell protein associated with immune signaling and cellular stress responses. Designed for sensitive, low‑background performance, it supports reliable detection in ELISA, Western blotting, and orthogonal analytical workflows used in bioprocess development. The antibody’s strong affinity enables accurate monitoring of chemokine impurities across upstream and downstream samples, aiding in process characterization, impurity clearance studies, and regulatory‑aligned HCP profiling. The 0.1 mg format is ideal for pilot assays, method optimization, and small‑volume workflows requiring consistent, reproducible detection of CHO‑specific chemokine contaminants.
Catalog No. 50-004-35325 Supplier Immunology Consultants Supplier No. GCC265ALYZ
This affinity‑purified goat antibody provides highly specific detection of CHO‑derived C‑C Motif Chemokine, an important host cell protein associated with immune signaling and cellular stress responses. Designed for sensitive, low‑background performance, it supports reliable detection in ELISA, Western blotting, and orthogonal analytical workflows used in bioprocess development. The antibody’s strong affinity enables accurate monitoring of chemokine impurities across upstream and downstream samples, aiding in process characterization, impurity clearance studies, and regulatory‑aligned HCP profiling. The 0.1 mg format is ideal for pilot assays, method optimization, and small‑volume workflows requiring consistent, reproducible detection of CHO‑specific chemokine contaminants.