Learn More
Immunology Consultants anti-Hamster (CHO) C-C Motif Chemokine Antibody - Affinity Purified
SDP
Supplier:  Immunology Consultants GCC265ALY
Encompass_Preferred
These affinity‑purified goat antibodies provide high‑specificity detection of CHO‑derived C‑C Motif Chemokine, a host cell protein relevant to bioprocess monitoring and impurity profiling. The antibodies perform reliably in ELISA, Western blotting, and HCP characterization workflows. The 0.5 mg format supports large‑scale assay development, while the 0.1 mg format is ideal for pilot studies and limited‑volume testing. Both deliver strong affinity, low background, and consistent lot‑to‑lot performance.
Catalog No. 50-004-35497