Learn More
Immunology Consultants anti-Hamster (CHO) Clusterin Antibody - Affinity Purified
SDP
Catalog No. 5000435299
Encompass_Preferred
This affinity‑purified anti‑Hamster (CHO) Clusterin antibody provides a high‑quality reagent for detecting this multifunctional glycoprotein, which plays roles in protein folding, stress response, and cellular homeostasis. Clusterin can appear as a CHO‑derived host cell protein, making it relevant for impurity profiling and bioprocess monitoring. Supplied at 0.1 mg, this antibody delivers strong specificity and consistent performance across ELISA, Western blotting, and related immunoassays. Its CHO‑targeted reactivity makes it a valuable tool for researchers assessing HCP clearance, studying CHO cell biology, or developing analytical methods for process characterization.
Catalog No. 50-004-35299 Supplier Immunology Consultants Supplier No. CCLS65ALYZ
This affinity‑purified anti‑Hamster (CHO) Clusterin antibody provides a high‑quality reagent for detecting this multifunctional glycoprotein, which plays roles in protein folding, stress response, and cellular homeostasis. Clusterin can appear as a CHO‑derived host cell protein, making it relevant for impurity profiling and bioprocess monitoring. Supplied at 0.1 mg, this antibody delivers strong specificity and consistent performance across ELISA, Western blotting, and related immunoassays. Its CHO‑targeted reactivity makes it a valuable tool for researchers assessing HCP clearance, studying CHO cell biology, or developing analytical methods for process characterization.