Learn More
Immunology Consultants anti-Hamster (CHO) Legumain Antibody - Affinity Purified
SDP
Catalog No. 5000435494
Encompass_Preferred
This affinity‑purified antibody targets hamster (CHO) Legumain, a lysosomal cysteine protease involved in protein processing, antigen presentation, and cellular stress responses. Produced in chicken and purified to high specificity, it provides reliable detection of Legumain across CHO‑derived lysates, purified preparations, and process‑related impurity panels. Its robust performance makes it well suited for bioprocess monitoring, host cell protein characterization, and development of quantitative immunoassays. The antibody’s avian origin minimizes background in mammalian systems, supporting sensitive detection in ELISA, Western blotting, and other analytical workflows requiring consistent lot‑to‑lot performance.
Catalog No. 50-004-35494 Supplier Immunology Consultants Supplier No. CLGM65ALYZ
This affinity‑purified antibody targets hamster (CHO) Legumain, a lysosomal cysteine protease involved in protein processing, antigen presentation, and cellular stress responses. Produced in chicken and purified to high specificity, it provides reliable detection of Legumain across CHO‑derived lysates, purified preparations, and process‑related impurity panels. Its robust performance makes it well suited for bioprocess monitoring, host cell protein characterization, and development of quantitative immunoassays. The antibody’s avian origin minimizes background in mammalian systems, supporting sensitive detection in ELISA, Western blotting, and other analytical workflows requiring consistent lot‑to‑lot performance.