Learn More
Eagle Biosciences Inc Anti-HAV IgM ELISA
SDP
Supplier:  Eagle Biosciences Inc E11
Encompass
Positive detection of antibodies directed against the Hepatitis A virus (anti-HAV) is evidence of immunity to Hepatitis-A virus (1). After natural infection with the Hepatitis A-Virus, neutralising antibodies appear at the same time of Anti-HAV of IgG-Class formation (1).
Catalog No. 50-187-8655